Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NACAD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554294
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554294 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554294 Supplier Invitrogen™ Supplier No. PA554294
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QESTATVTPH TLQVAPGLQV EVATRVTPQA GEEETDSTAG QESAAMAMPQ PSQEGISEIL GQESVTAEKL PTPQEETSLT LCPDSPQNLK EEG Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 48%.

May prevent inappropriate targeting of non-secretory polypeptides to the endoplasmic reticulum (ER). May bind to nascent polypeptide chains as they emerge from the ribosome and block their interaction with the signal recognition particle (SRP), which normally targets nascent secretory peptides to the ER. May also reduce the inherent affinity of ribosomes for protein translocation sites in the ER membrane (M sites).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NACAD
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NACAD
Gene Accession No. O15069
Gene Alias D230024G13Rik; Kiaa0363; mKIAA0363; NAC alpha domain containing; NACAD; NAC-alpha domain-containing protein 1
Gene Symbols NACAD
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NACAD. Recombinant protein control fragment (Product #RP-92733).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23148
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.