Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NACAD Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP31712925UL
This item is not returnable.
View return policy
Description
NACAD Polyclonal antibody specifically detects NACAD in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
NACAD | |
Polyclonal | |
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
KIAA0363NAC-alpha domain-containing protein 1, NAC alpha domain containing | |
This antibody was developed against Recombinant Protein corresponding to amino acids: DQVQQDDPQPAAEAGTPWAAQEDADSTLGMEALSLPEPASGAGEEIAEALSRPGREACLEARAHTGDGAKPDSPQKETLE | |
25 μg | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
23148 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction