Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NACC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561624
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561624 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561624 Supplier Invitrogen™ Supplier No. PA561624
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RVRKRWLPKI KSMLPEGVEM YRTVMGSAAA SVPLDPEFPP AAAQVFEQRI YAERRGDAAT IVALRTDAVN VDLSAAAN Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

Functions as a transcriptional repressor through its association with the NuRD complex. Recruits the NuRD complex to the promoter of MDM2, leading to the repression of MDM2 transcription and subsequent stability of p53/TP53. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen NACC2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Nacc2
Gene Accession No. Q96BF6
Gene Alias 0610020I02Rik; AI448087; BEN domain containing 9; BEND9; BTB (POZ) domain containing 14A; BTB/POZ domain-containing protein 14A; BTBD14; Btbd14a; BTBD31; C030048H19Rik; MGC23427; NAC2; NAC-2; NACC family member 2; NACC family member 2, BEN and BTB (POZ) domain containing; Nacc2; nucleus accumbens associated 2, BEN and BTB (POZ) domain containing; nucleus accumbens-associated protein 2; RBB; repressor with BTB domain and BEN domain; transcription repressor with a BTB domain and a BEN domain
Gene Symbols Nacc2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NACC2. Recombinant protein control fragment (Product #RP-100971).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 138151
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.