Learn More
Invitrogen™ NADE Polyclonal Antibody

Description
Despite its predicted molecular weight, NADE often migrates at ~23kDa in SDS-PAGE. A suggested positive control is human brain tissue lysate. PA5-20076 can be used with blocking peptide PEP-0194.
Specifications
Specifications
| Antigen | NADE |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | BEX3 |
| Gene Accession No. | Q00994, Q9WTZ9 |
| Gene Alias | AL033356; Bex; Bex3; brain expressed X-linked 3; brain-expressed X-linked protein 3; Brain-expressed X-linked protein 3 homolog; DXS6984E; DXWsu67e; Gcap27; granule cell antiserum positive 27; HGR74; Nade; nerve growth factor receptor (TNFRSF16) associated protein 1; nerve growth factor receptor associated protein 1; nerve growth factor receptor-associated protein 1; Ngfrap1; ovarian granulosa cell 13.0 kDa protein HGR74; p75NTR-associated cell death executor; protein BEX3; rBex3; RP13-349O20.1 |
| Gene Symbols | BEX3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.