Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NADE Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA527690
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (80%), Pig (95%), Rhesus Monkey (100%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May be a signaling adapter molecule involved in p75NTR-mediated apoptosis induced by NGF. Plays a role in zinc-triggered neuronal death. May play an important role in the pathogenesis of neurogenetic diseases.
Specifications
| NADE | |
| Polyclonal | |
| Unconjugated | |
| BEX3 | |
| AL033356; Bex; Bex3; brain expressed X-linked 3; brain-expressed X-linked protein 3; Brain-expressed X-linked protein 3 homolog; DXS6984E; DXWsu67e; Gcap27; granule cell antiserum positive 27; HGR74; Nade; nerve growth factor receptor (TNFRSF16) associated protein 1; nerve growth factor receptor associated protein 1; nerve growth factor receptor-associated protein 1; Ngfrap1; ovarian granulosa cell 13.0 kDa protein HGR74; p75NTR-associated cell death executor; protein BEX3; rBex3; RP13-349O20.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27018 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.24 mg/mL | |
| PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q00994 | |
| BEX3 | |
| Recombinant fragment corresponding to a region within amino acids 1 and 111 of Human NADE. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction