Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NADE Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110029
Description
Immunogen sequence: MANIHQENEE MEQPMQNGEE DRPLGGGEGH QPAGNRRGQA RRLAPNFRWA IPNRQINDGM GGDGDDMEIF MEEMREIRRK LRELQLRNCL RILMGELSNH HDHHDEFCLM P.
May be a signaling adapter molecule involved in p75NTR-mediated apoptosis induced by NGF. Plays a role in zinc-triggered neuronal death. May play an important role in the pathogenesis of neurogenetic diseases.
Specifications
| NADE | |
| Polyclonal | |
| Unconjugated | |
| BEX3 | |
| AL033356; Bex; Bex3; brain expressed X-linked 3; brain-expressed X-linked protein 3; Brain-expressed X-linked protein 3 homolog; DXS6984E; DXWsu67e; Gcap27; granule cell antiserum positive 27; HGR74; Nade; nerve growth factor receptor (TNFRSF16) associated protein 1; nerve growth factor receptor associated protein 1; nerve growth factor receptor-associated protein 1; Ngfrap1; ovarian granulosa cell 13.0 kDa protein HGR74; p75NTR-associated cell death executor; protein BEX3; rBex3; RP13-349O20.1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12070, 27018 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q00994, Q9WTZ9 | |
| BEX3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-111 of human NGFRAP1 (NP_996798.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction