Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NADK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570657
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570657 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570657 Supplier Invitrogen™ Supplier No. PA570657
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

NADK (Nicotinamide Adenine Dinucleotide Kinase) is an enzyme crucial in the regulation of intracellular NADP+ levels, which are necessary for metabolic processes such as fatty acid and DNA synthesis. The human NADK gene is located on chromosome 1p36.33 and encodes a protein that primarily functions in the cytoplasm. NADK catalyzes the phosphorylation of NAD+ to NADP+, playing a significant role in maintaining the cellular redox state. Structurally, human NADK forms a dimeric organization, as revealed by its crystal structure, and includes four subunits (chains A-D) with specific colors indicating different chains. The enzyme's activity is regulated post-translationally, but its pathophysiological significance in humans remains underexplored. Multiple orthologs of the NADK gene are present in various organisms, illustrating its evolutionary conservation and importance across different species.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NADK
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NADK
Gene Accession No. O95544
Gene Alias 4432404C02Rik; BC004012; dJ283E3.1; FLJ13052; hypothetical protein LOC513419; LOW QUALITY PROTEIN: NAD kinase; NAD kinase; Nadk; Poly(P)/ATP NAD kinase; ppnk; Putative inorganic polyphosphate/ATP-NAD kinase (Poly(P)/ATP NAD kinase)
Gene Symbols NADK
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human NADK (aa 374-423).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65220
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.