Learn More
NADK Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304993AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
NADK catalyzes the transfer of a phosphate group from ATP to NAD to generate NADP, which in its reduced form acts as an electron donor for biosynthetic reactions.Specifications
NADK | |
Polyclonal | |
Unconjugated | |
NADK | |
dJ283E3.1, FLJ13052, poly(P)/ATP NAD kinase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
192185, 65220 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
O95544, P58058 | |
NADK | |
Between 25 and 75 | |
10 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.