Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ NADPH reduced form, tetrasodium salt
GREENER_CHOICESDP

Catalog No. 5515100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5515100 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 5515100 Supplier R&D Systems™ Supplier No. 5515/100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Cofactor for many anabolic enzymes

NADPH reduced form tetrasodium salt is a cofactor for many anabolic enzymes which use NADPH as a reducing agent.

Specifications

Chemical Name or Material NADPH reduced form tetrasodium salt
CAS 2646-71-1
Synonym β-Nicotinamide adenine dinucleotide 2'-phosphate reduced tetrasodium salt
Quantity 100 mg
Solubility Soluble to 50mM in water
Product Line Tocris
Molecular Formula C21H26N7Na4O17P3
Formula Weight 833.35
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >93%
Show More Show Less

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.