Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NADSYN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565213
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565213 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565213 Supplier Invitrogen™ Supplier No. PA565213
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VRSGNEEVLAD VRTIVNQISY TPQDPRDLCG RILTTCYMAS KNSSQETCTR ARELAQQIGS HHISLNIDPA VKAVMGIFSL VTGKSPLFAA HGGSSRENLA LQNVQARIRM VL Highest antigen sequence identity to the following orthologs - mouse 76%, rat 80%.

Nicotinamide adenine dinucleotide (NAD) is a coenzyme in metabolic redox reactions, a precursor for several cell signaling molecules, and a substrate for protein posttranslational modifications. NAD synthetase (EC 6.3.5.1) catalyzes the final step in the biosynthesis of NAD from nicotinic acid adenine dinucleotide (NaAD).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NADSYN1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NADSYN1
Gene Accession No. Q6IA69
Gene Alias glutamine-dependent NAD synthetase; glutamine-dependent NAD(+) synthetase; NAD synthetase 1; NAD(+) synthase; NAD(+) synthase [glutamine-hydrolyzing]; NAD(+) synthetase; NADSYN1
Gene Symbols NADSYN1
Host Species Rabbit
Immunogen Recombinant Human NADSYN1. Recombinant protein control fragment (Product #RP-104689).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55191
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.