Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NAE1 Monoclonal Antibody (OTI1E10)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526665
Description
NAE1 Monoclonal Antibody for Western Blot, IHC (P)
The protein encoded by this gene binds to the beta-amyloid precursor protein. Beta-amyloid precursor protein is a cell surface protein with signal-transducing properties, and it is thought to play a role in the pathogenesis of Alzheimer's disease. In addition, the encoded protein can form a heterodimer with UBE1C and bind and activate NEDD8, a ubiquitin-like protein. This protein is required for cell cycle progression through the S/M checkpoint. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
| NAE1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13564 | |
| NAE1 | |
| Human recombinant protein fragment corresponding to amino acids 1-274 of NAE1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1E10 | |
| Unconjugated | |
| NAE1 | |
| 59kDa; A-116A10.1; amyloid beta precursor protein binding protein 1, 59kDa; amyloid beta precursor protein-binding protein 1, 59 kDa; amyloid beta precursor protein-binding protein 1, 59kD; amyloid protein-binding protein 1; Appbp1; APP-BP1; HPP1; Nae1; NEDD8 activating enzyme E1 subunit 1; NEDD8-activating enzyme E1 regulatory subunit; NEDD8-activating enzyme E1 subunit; protooncogene protein 1; proto-oncogene protein 1; ula-1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8883 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction