Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAF1 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP222263AF350
Description
NAF1 Polyclonal antibody specifically detects NAF1 in Human samples. It is validated for Western BlotSpecifications
NAF1 | |
Polyclonal | |
Western Blot | |
nuclear assembly factor 1 homolog (S. cerevisiae) | |
The immunogen this antibody was made to, maps to a region between residue 50 and 100 of human Nuclear Assembly Factor 1 using the numbering given in entry NP_612395.2 (GeneID 92345). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
92345 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction