Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116774
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116774 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116774 Supplier Invitrogen™ Supplier No. PA5116774
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: K-562, Mouse testis Immunogen sequence: PPVLSDGDDD LQIEKENKNF PLKTKDELLL NELPSVEELT IILPEDIELK PLGMVSSIIE QLVIIESMTN LPPVNEETVI FKSDRQAAGK IFEIFGPVAH PFYVLRFNSS DHIESKGIKI KETMYFAPSM KDFTQYIFTE KLKQDKGSDA SWKNDQEPPP EALDFSDDEK EKEAKQRKKS QIQGRKKLKS EFNEPGEDFT EVHQNWNAHS SASEHAKGYR NREFTRGFSR ARYPRSCHGR PPPQHFYNSE H.

RNA-binding protein required for the maturation of box H/ACA snoRNPs complex and ribosome biogenesis. During assembly of the H/ACA snoRNPs complex, it associates with the complex and disappears during maturation of the complex and is replaced by NOLA1/GAR1 to yield mature H/ACA snoRNPs complex. Probably competes with NOLA1/GAR1 for binding with DKC1/NOLA4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAF1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NAF1
Gene Accession No. Q3UMQ8, Q52KK4, Q96HR8
Gene Alias Gm174; H/ACA ribonucleoprotein complex non-core subunit NAF1; hNAF1; NAF1; nuclear assembly factor 1 homolog; nuclear assembly factor 1 homolog (S. cerevisiae); nuclear assembly factor 1 ribonucleoprotein; RGD1306802
Gene Symbols NAF1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 150-400 of human NAF1 (NP_6123952).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 234344, 306387, 92345
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.