Learn More
NAF1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303911A

Description
The recommended shelf life for this product is 1 year from date of receipt.
RNA-binding protein required for the maturation of box H/ACA snoRNPs complex and ribosome biogenesis. During assembly of the H/ACA snoRNPs complex, it associates with the complex and disappears during maturation of the complex and is replaced by NOLA1/GAR1 to yield mature H/ACA snoRNPs complex. Probably competes with NOLA1/GAR1 for binding with DKC1/NOLA4.Specifications
NAF1 | |
Polyclonal | |
Unconjugated | |
NAF1 | |
hNAF1, nuclear assembly factor 1 homolog, nuclear assembly factor 1 homolog (S. cerevisiae) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
92345 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q96HR8 | |
NAF1 | |
Between 50 and 100 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.