Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAG-1/GDF15 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 07217IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-217-IMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-217-IMI Supplier MilliporeSigma Supplier No. 07217I
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Growth/differentiation factor 15 using this rabbit polyclonal Anti-NAG-1/GDF15 useing Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAG-1/GDF15
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols GDF15,MIC1,PDF,PLAB,PTGFB
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 16 amino acids from the C-terminal end of human NAG-1/GDF15.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_004855
Test Specificity This polyclonal antibody specifically detects Growth/differentiation factor 15 in human.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.