Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAG-1 Monoclonal Antibody (6D12.H10.E4), Rockland™
SDP

Catalog No. 501943812
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-194-3812 100 μg
50-199-1226 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-194-3812 Supplier Rockland Immunochemicals Supplier No. 200501B07
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody reacts with the C-terminus of endogenous NAG-1 protein from mouse tissues. A BLAST analysis suggests reactivity with NAG-1 from chimpanzee and macaque based on a 100% homology. Partial reactivity is expected against rat based on an 86% homology with the immunizing sequence. Cross-reactivity with NAG-1 from other sources has not been determined.

Bone morphogenetic proteins (e. g., BMP5, MIM 112265) are members of the transforming growth factor-beta (see TGFB1, MIM 190180) superfamily and regulate tissue differentiation and maintenance. They are synthesized as precursor molecules that are processed at a dibasic cleavage site to release C-terminal domains containing a characteristic motif of 7 conserved cysteines in the mature protein.

Specifications

Antigen NAG-1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 6D12.H10.E4
Concentration 1.09 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene Gdf15
Gene Accession No. Q99988, Q9Z0J7
Gene Alias GDF15; GDF-15; growth differentiation factor 15; Growth/differentiation factor 15; macrophage inhibiting cytokine-1; Macrophage inhibitory cytokine 1; MIC1; MIC-1; NAG1; NAG-1; NRG-1; NSAID (nonsteroidal anti-inflammatory drug)-activated protein 1; NSAID-activated gene 1 protein; NSAID-regulated gene 1 protein; PDF; PLAB; Placental bone morphogenetic protein; Placental TGF-beta; Prostate differentiation factor; PTGFB; PTGF-beta; Sbf
Gene Symbols Gdf15
Host Species Rat
Immunogen This Protein-A purified antibody was prepared by repeated immunizations with an MBP-tagged recombinant protein produced in E.coli corresponding to C-term mouse NAG-1 protein. Cross reactivity to MBP is negative.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23886, 9518
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.