Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAG Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318441JF646
Description
NAG Polyclonal antibody specifically detects NAG in Human samples. It is validated for Western BlotSpecifications
NAG | |
Polyclonal | |
Western Blot | |
DKFZp586G1219, EC 1.1.1.94, EC 1.2.1.59, FLJ40407, NAG/BC035112 fusion, NAG/FAM49A fusion, NAGneuroblastoma-amplified protein, neuroblastoma amplified sequence, Neuroblastoma-amplified gene protein, neuroblastoma-amplified sequence | |
Recognizes region between residues Between Between 350 and 400.. | |
0.1 mL | |
Cell Biology | |
51594 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction