Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAGK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557195
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557195 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557195 Supplier Invitrogen™ Supplier No. PA557195
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SEVLLVSEDG KILAEADGLS TNHWLIGTDK CVERINEMVN RAKRKAGVDP LVPLRSLGLS LSGGDQEDAG RILIEELRDR FPYLSE Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 88%.

SPG20 is a protein containing a MIT (Microtubule Interacting and Trafficking molecule) domain, and is implicated in regulating endosomal trafficking and mitochondria function. The protein localizes to mitochondria and partially co-localizes with microtubules. Stimulation with epidermal growth factor (EGF) results in protein translocation to the plasma membrane, and the protein functions in the degradation and intracellular trafficking of EGF receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAGK
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NAGK
Gene Accession No. Q9UJ70
Gene Alias GlcNAc kinase; Gnk; HSA242910; N-acetyl-D-glucosamine kinase; N-acetylglucosamine kinase; Nagk
Gene Symbols NAGK
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NAGK. Recombinant protein control fragment (Product #RP-102655).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55577
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.