Learn More
NAGK Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302680AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
N-acetylglucosamine kinase (NAGK) converts endogenous N-acetylglucosamine (GlcNAc), a major component of complex carbohydrates, from lysosomal degradation or nutritional sources into GlcNAc 6-phosphate. NAGK belongs to the group of N-acetylhexosamine kinases and is a prominent salvage enzyme of amino sugar metabolism in mammals. The predicted 344-amino acid NAGK protein contains the 5 sequence motifs necessary for the binding of ATP by sugar kinases. NAGK shares 91.6% amino acid similarity with mouse Nagk, for which enzyme activity is detectable in all mouse tissues examined, with highest enzymatic activity in testis. It is hypothesized that NAGK has a general role in the catabolic pathways of GlcNAc as well as of ManNAc.Specifications
NAGK | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9UJ70 | |
NAGK | |
Between 50 and 100 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
NAGK | |
GNK, HSA242910 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55577 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.