Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAGK Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501563200
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-3200 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-3200 Supplier Bethyl Laboratories, Inc Supplier No. A302680A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

N-acetylglucosamine kinase (NAGK) converts endogenous N-acetylglucosamine (GlcNAc), a major component of complex carbohydrates, from lysosomal degradation or nutritional sources into GlcNAc 6-phosphate. NAGK belongs to the group of N-acetylhexosamine kinases and is a prominent salvage enzyme of amino sugar metabolism in mammals. The predicted 344-amino acid NAGK protein contains the 5 sequence motifs necessary for the binding of ATP by sugar kinases. NAGK shares 91.6% amino acid similarity with mouse Nagk, for which enzyme activity is detectable in all mouse tissues examined, with highest enzymatic activity in testis. It is hypothesized that NAGK has a general role in the catabolic pathways of GlcNAc as well as of ManNAc.

Specifications

Antigen NAGK
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene NAGK
Gene Accession No. Q9UJ70
Gene Alias GNK, HSA242910
Gene Symbols NAGK
Host Species Rabbit
Immunogen Between 50 and 100
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55577
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.