Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NAGLU Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | NAGLU |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119299
|
Invitrogen™
PA5119299 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121749
|
Invitrogen™
PA5121749 |
20 μL |
Each for $209.50
|
|
|||||
Description
NAGLU Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes an enzyme that degrades heparan sulfate by hydrolysis of terminal N-acetyl-D-glucosamine residues in N-acetyl-alpha-D-glucosaminides. Defects in this gene are the cause of mucopolysaccharidosis type IIIB (MPS-IIIB), also known as Sanfilippo syndrome B. This disease is characterized by the lysosomal accumulation and urinary excretion of heparan sulfate.Specifications
NAGLU | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human, Pig | |
NAGLU | |
Alpha-N-acetylglucosaminidase; alpha-N-acetylglucosaminidase (Sanfilippo disease IIIB); Alpha-N-acetylglucosaminidase 77 kDa form; Alpha-N-acetylglucosaminidase 82 kDa form; alpha-N-acetylglucosaminidase, lysosomal; CMT2V; MPS3B; MPS-IIIB; N-acetyl-alpha-glucosaminidase; N-acetyl-glucosaminidase; N-acetylglucosaminidase, alpha; NAG; NAGLU; RGD1564228; testicular tissue protein Li 18; UFHSD; UFHSD1 | |
NAGLU | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
P54802 | |
100519685, 4669 | |
Recombinant protein N-Acetyl Alpha-D-Glucosaminidase. The antigen corresponds to amino acid range 485-743 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title