Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAGPA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564149
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564149 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564149 Supplier Invitrogen™ Supplier No. PA564149
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FFRMNSGECL GNVVSDERRV SSSGGLQNAQ FGIRRDGTLV TGYLSEEEVL DTENPFVQLL SGVVWLIRNG SIYINESQAT ECDETQETGS FSKFVNVISA RTAIGHDRKG QLVLFHADGQ TEQRGINLWE MAE Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 92%.

Hydrolases are transported to lysosomes after binding to mannose 6-phosphate receptors in the trans-Golgi network. This gene encodes the enzyme that catalyzes the second step in the formation of the mannose 6-phosphate recognition marker on lysosomal hydrolases. Commonly known as 'uncovering enzyme' or UCE, this enzyme removes N-acetyl-D-glucosamine (GlcNAc) residues from GlcNAc-alpha-P-mannose moieties and thereby produces the recognition marker. The encoded preproprotein is proteolytically processed by furin to generate the mature enzyme, a homotetramer of two disulfide-linked homodimers. Mutations in this gene are associated with developmental stuttering in human patients.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAGPA
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NAGPA
Gene Accession No. Q9UK23
Gene Alias AI596180; alpha-GlcNAcase; alpha-N-acetylglucosaminyl phosphodiesterase; APAA; lysosomal alpha-N-acetylglucosaminidase; mannose 6-phosphate-uncovering enzyme; N-acetylglucosamine-1-phosphodiester alpha-N-acetylglucosaminidase; NAGPA; Phosphodiester alpha-GlcNAcase; UCE; zgc:165534
Gene Symbols NAGPA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NAGPA. Recombinant protein control fragment (Product #RP-103542).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51172
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.