Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAGS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565010
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565010 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565010 Supplier Invitrogen™ Supplier No. PA565010
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PTKIIFLNNTG GLRDSSHKVL SNVNLPADLD LVCNAEWVST KERQQMRLIV DVLSRLPHHS SAVITAASTL LTELFSNKGS GTLFKNAERM LRVRSLDKLD QGRLVDLV Highest antigen sequence identity to the following orthologs - mouse 87%, rat 90%.

NAGS is a mitochondrial enzyme that catalyzes the formation of N-acetylglutamate (NAG) from glutamate and acetyl coenzyme-A. NAG is a cofactor of carbamyl phosphate synthetase I (CPSI), the first enzyme of the urea cycle in mammals. Deficiencies in N-acetylglutamate synthase have been associated with hyperammonemia.The N-acetylglutamate synthase gene encodes a mitochondrial enzyme that catalyzes the formation of N-acetylglutamate (NAG) from glutamate and acetyl coenzyme-A. NAG is a cofactor of carbamyl phosphate synthetase I (CPSI), the first enzyme of the urea cycle in mammals. This gene may regulate ureagenesis by altering NAG availability and, thereby, CPSI activity. Deficiencies in N-acetylglutamate synthase have been associated with hyperammonemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAGS
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NAGS
Gene Accession No. Q8N159
Gene Alias 1700120E20Rik; AGAS; Amino-acid acetyltransferase; amino-acid N-acetyltransferase; ARGA; LOW QUALITY PROTEIN: N-acetylglutamate synthase, mitochondrial; MGC133025; N-acetylglutamate synthase; N-acetylglutamate synthase conserved domain form; N-acetylglutamate synthase long form; N-acetylglutamate synthase short form; N-acetylglutamate synthase, conserved domain form; N-acetylglutamate synthase, mature form; N-acetylglutamate synthase, mitochondrial; Nags; NAGS-C; NAGS-M; RGD1565783
Gene Symbols NAGS
Host Species Rabbit
Immunogen Recombinant Human NAGS. Recombinant protein control fragment (Product #RP-105775).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 162417
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.