Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAIF1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550241

Catalog No. PA550241


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total NAIF1 protein.

Human NAIF1 is located on chromosome 9q34.11 and encodes 327 amino acids with a homeodomain-like region and two nuclear localization signals at its N-terminal region. NAIF1 is conserved across diverse species, including human, mouse, crab-eating macaque, dog, chicken and frog, and shares no obvious homology to any known genes or proteins. NAIF1 was predominantly localized in the nucleus. Overexpression of NAIF1 inhibited cell growth and induced apoptosis. Furthermore, NAIF1 transfection caused both decreases in mitochondrial membrane potential and caspase-3 activation. In summary, NAIF1 is a nuclear protein that induces apoptosis when overexpressed.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NAIF1
Polyclonal
Unconjugated
NAIF1
2310007O20Rik; 4933440H19Rik; bA379C10.2; C9orf90; NAIF1; nuclear apoptosis inducing factor 1; nuclear apoptosis-inducing factor 1
Rabbit
Antigen Affinity Chromatography
RUO
203245
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1.5 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q69YI7
NAIF1
Full length fusion protein of Human NAIF1.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.