Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAIP Antibody (SR1227), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | NAIP |
---|---|
Clone | SR1227 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
NAIP Monoclonal antibody specifically detects NAIP in Human samples. It is validated for Western BlotSpecifications
NAIP | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
baculoviral IAP repeat-containing 1, BIRC1baculoviral IAP repeat-containing protein 1, FLJ18088, FLJ42520, FLJ58811, Neuronal apoptosis inhibitory protein, NLR family, apoptosis inhibitory protein, NLR family, BIR domain containing 1, NLRB1, nucleotide-binding oligomerization domain, leucine rich repeat and BIR domaincontaining 1, psi neuronal apoptosis inhibitory protein, psiNAIP | |
A synthesized peptide derived from human NAIP (Uniprot #: Q13075) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1227 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
4671 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title