Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115614
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115614 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115614 Supplier Invitrogen™ Supplier No. PA5115614
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NAIP.

Neuronal apoptosis inhibitor protein (NAIP) was the first human inhibitor of apoptosis protein (IAP) identified and was discovered by its association with the neurodegenerative disorder spinal muscular atrophy. Members of the IAP family contain one to three copies of an approximately 70 amino acid motif termed baculovirus IAP repeat (BIR); these BIRs promote protein-protein interactions with various caspases such as caspase-3, -7, and -9 as well as members of the TRAF family of signal molecules (reviewed in 3). Unlike other IAPs however, NAIP requires ATP to bind caspase-9 and is not inhibited by the IAP-inhibiting molecule Smac/DIABLO, suggesting that NAIP is unique among the IAPs in its regulation of its activity. Finally, although only one human NAIP protein has been identified, other shorter NAIP mRNA transcripts have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAIP
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene NAIP
Gene Accession No. Q13075, Q9QWK5
Gene Alias AV364616; baculoviral IAP repeat-containing 1a; baculoviral IAP repeat-containing protein 1; Baculoviral IAP repeat-containing protein 1a; BIRC1; Birc1a; D13Lsd1; FLJ18088; FLJ42520; FLJ58811; Naip; Naip1; Naip-rs1; Neuronal apoptosis inhibitory protein; neuronal apoptosis inhibitory protein 1; NLR family apoptosis inhibitory protein; NLR family, apoptosis inhibitory protein; NLR family, apoptosis inhibitory protein 1; NLRB1; nucleotide-binding oligomerization domain, leucine rich repeat and BIR domain containing 1; psi neuronal apoptosis inhibitory protein; psiNAIP
Gene Symbols NAIP, Naip1
Host Species Rabbit
Immunogen A synthesized peptide derived from human NAIP(Accession Q13075), corresponding to amino acid residues A302-A352.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17940, 4671
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.