Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naked1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989867
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9867 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9867 Supplier Bioss Supplier No. BS19005R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

In the mouse, Nkd is a Dishevelled (see DVL1; MIM 601365)-binding protein that functions as a negative regulator of the Wnt (see WNT1; MIM 164820)-beta-catenin-Tcf signaling pathway.

Specifications

Antigen Naked1
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene NKD1
Gene Accession No. Q969G9
Gene Alias 2810434J10Rik; 9030215G15Rik; Dvl-binding protein; hNkd; hNkd1; mNkd; mNkd1; naked cuticle 1 homolog; naked cuticle 1 homolog (Drosophila); naked cuticle homolog 1; naked cuticle homolog 1 (Drosophila); naked cuticle-1; Naked1; Naked-1; NKD; Nkd1; PP7246; Protein naked cuticle homolog 1
Gene Symbols NKD1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Naked1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85407
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.