Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Naked2 Monoclonal Antibody (S.902.7)
GREENER_CHOICE

Catalog No. PIMA514827
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514827 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514827 Supplier Invitrogen™ Supplier No. MA514827
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

It is not recommended to aliquot this antibody.

Cell autonomous antagonist of the canonical Wnt signaling pathway. May activate a second Wnt signaling pathway that controls planar cell polarity. Required for processing of TGFA and for targeting of TGFA to the basolateral membrane of polarized epithelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Naked2
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone S.902.7
Conjugate Unconjugated
Formulation 0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5
Gene Nkd2
Gene Accession No. Q969F2
Gene Alias 2210403L10Rik; AW212591; Dvl-binding protein NKD2; hNkd2; mNkd2; naked cuticle 1 homolog; naked cuticle 2 homolog; naked cuticle 2 homolog (Drosophila); naked cuticle homolog 2; naked cuticle homolog 2 (Drosophila); naked cuticle-2; Naked2; naked-2; Nkd2; Protein naked cuticle homolog 2
Gene Symbols Nkd2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues surrounding Ala217 of human Naked2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85409
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.