Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Naked2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA561979
Description
Immunogen sequence: GHKRYRQKGR EGHSPLKAPH AQPATVEHEV VRDLPPTPAG EGYAVPVIQR HE Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 77%.
Cell autonomous antagonist of the canonical Wnt signaling pathway. May activate a second Wnt signaling pathway that controls planar cell polarity. Required for processing of TGFA and for targeting of TGFA to the basolateral membrane of polarized epithelial cells.
Specifications
Naked2 | |
Polyclonal | |
Unconjugated | |
Nkd2 | |
2210403L10Rik; AW212591; Dvl-binding protein NKD2; hNkd2; mNkd2; naked cuticle 1 homolog; naked cuticle 2 homolog; naked cuticle 2 homolog (Drosophila); naked cuticle homolog 2; naked cuticle homolog 2 (Drosophila); naked cuticle-2; Naked2; naked-2; Nkd2; Protein naked cuticle homolog 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
85409 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
1.0 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q969F2 | |
Nkd2 | |
Recombinant protein corresponding to Human Naked2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction