Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NALCN Monoclonal Antibody (N187/7), PerCP
GREENER_CHOICE

Catalog No. PIMA545574
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545574 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545574 Supplier Invitrogen™ Supplier No. MA545574
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45574 was sufficient for detection of NALCN in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 200kDa. This antibody was formerly sold as clone S187-7.

Nalcn is a voltage-independent, cation-nonselective channel which is permeable to sodium, potassium and calcium ions. Nalcn regulates the resting membrane potential and controls neuronal excitability (PubMed:17448995). Neuropeptides such as neurotensin and substance P (SP) stimulate the firing of action potentials by activating NALCN through a SRC family kinases-dependent pathway. In addition to its baseline activity, Nalcn activity is enhanced/modulated by several GPCRs, is required for normal respiratory rhythm and neonatal survival, is involved in systemic osmoregulation by controlling the serum sodium concentration. Nalcn is partly responsible for the substance P-induced depolarization and regulation of the intestinal pace-making activity in the interstitial cells of Cajal. Further, Nalcn plays a critical role in both maintenance of spontaneous firing of substantia nigra pars reticulata (SNr) neurons and physiological modulation of SNr neuron excitability. Diseases associated with NALCN include Congenital contractures of the limbs and face, Hypotonia, Developmental Delay and Hypotonia, Infantile Psychomotor Retardation And Characteristic Facies 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NALCN
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N187/7
Concentration 1 mg/mL
Conjugate PerCP
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene NALCN
Gene Accession No. Q6Q760, Q8BXR5, Q8IZF0
Gene Alias A530023G15Rik; AI849508; bA430M15.1; brain voltage-gated cation channel; CanIon; CLIFAHDD; FLJ23913; FLJ44659; FLJ44764; Four domain-type voltage-gated ion channel alpha-1 subunit; four repeat voltage-gated ion channel; IHPRF; IHPRF1; INNFD; MGC74524; Nalcn; Nca; Rb21; Rb21-channel; Sodium leak channel non-selective protein; sodium leak channel, non selective; sodium leak channel, non-selective; Vgcnl1; voltage gated channel like 1; voltage gated channel-like protein 1
Gene Symbols NALCN
Host Species Mouse
Immunogen Fusion protein amino acids 1659-1738 (cytoplasmic C-terminus) of rat NALCN.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259232, 266760, 338370
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.