Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Syringe Filters
GREENER_CHOICE

Catalog No. 09740100
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
0.8 μm
Diameter (Metric):
4 mm
13 mm
25 mm
Material (Membrane):
Cellulose Acetate
Glass Fiber, Cellulose Acetate
Nylon
PES
SFCA
Sterility:
Non-sterile
Sterile
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Diameter (Metric) Material (Membrane) Sterility
09-740-100 0.2 μm 13 mm PES Sterile
09-740-104 0.2 μm 25 mm Glass Fiber, Cellulose Acetate Sterile
09-740-105 0.2 μm 25 mm SFCA Sterile
09-740-106 0.45 μm 25 mm SFCA Sterile
09-740-107 0.8 μm 25 mm Cellulose Acetate Sterile
09-740-108 0.2 μm 25 mm SFCA Sterile
09-740-109 0.45 μm 25 mm SFCA Sterile
09-740-113 0.2 μm 25 mm PES Sterile
09-740-114 0.45 μm 25 mm PES Sterile
09-740-115 0.2 μm 25 mm Nylon Sterile
09-740-116 0.45 μm 25 mm Nylon Sterile
09-740-34A 0.2 μm 4 mm Cellulose Acetate Non-sterile
09-740-34B 0.45 μm 4 mm Cellulose Acetate Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 09-740-100 Supplier Thermo Scientific™ Supplier No. 7201320
Add to Cart
Add to Cart

Filter sample volumes of 10 to 100mL with these sterile syringe filters featuring a variety of membranes and pore sizes for most laboratory applications.

Thermo Scientific™ Nalgene™ syringe filters are built to meet your lab’s clarification and sterilization needs. Our filters are available in a variety of sizes and membranes that can be easily used for both sterile and non-sterile laboratory applications. Whether your research necessitates prefiltration, sample preparation, sterile filtration, laboratory filtration, or gas venting, Nalgene syringe filters are lab essentials.

Key Features
  • 10 to 100mL sample size
  • Inlet: Female Luer-Lok™
  • Outlet: Male Luer slip
  • Polypropylene housing with ID color rings
  • Pore size and membrane type printed on unit

Surfactant-free Cellulose Acetate (SFCA)

  • Low protein binding, lower extractables than standard CA
  • Good for cell culture fluids and cellular solutions
  • Polypropylene housings with blue color rings

Cellulose Acetate (CA)

  • Some protein binding and extractables
  • For sterile filtration and clarification of: buffers, aqueous solutions and reagents
  • Polypropylene housings with red color rings

Nylon

  • Naturally hydrophilic, low extractables, some protein binding
  • Useful for alcohols, cryoprotectants such as DMSO and with aqueous/biological and weak solvents
  • Polypropylene housings with pink color rings
PTFE
  • Naturally hydrophobic
  • Excellent for HPLC/GC solvent filtration. Ideal for sterile venting of air or other gases during filtration
  • Polypropylene housings with white color rings

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Certified sterile, non-cytotoxic and non-pyrogenic
Filter Type Membrane Filter
Material (Membrane) PES
Pore Size 0.2 μm
Sterility Sterile
For Use With (Application) Sterile filtration and clarification of cell culture media, buffers and additives
Housing Material Polypropylene
Quantity 100/Pk.
Type Syringe Filter
Volume (Metric) 2 to 10 mL
Diameter (Metric) 13 mm
Show More Show Less
Are Nalgene Sterile Syringe Filters packed individually or in bulk?

The Nalgene Sterile Syringe Filters are individually blister-packed. The Nalgene non-Sterile Syringe Filters are bulk packed.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.