Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ PC Platinum Certified Clean™ Biotainer™ Bottle
GREENER_CHOICE

Catalog No. 14388038
Click to view available options
Capacity (Metric):
5 mL
20 mL
1 L
2 L
5 L
10 L
Shape:
Round
Square

Thermo Scientific™ Nalgene™ PC Platinum Certified Clean™ Biotainer™ Bottles are certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination.

Products are washed with USP Water for Injection and handled post-washing in Class 5 clean rooms with Class 4 packaging/inspection hoods to produce these very low particulate products. Certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination. Sterile, ready-to-use products with a Sterility Assurance Level of 10-6 inside the container/closure, including two heat-sealed sterile barriers to protect product prior to use.

  • Reduce the risk of carry-over contamination and eliminate the cost of cleaning, sterilizing and associated validations
  • Durable, translucent PC construction ensures the integrity of your sample
  • Designed specifically for pharmaceutical and biological manufacturers
  • Closures feature a silicone liner for leakproof assurance
  • Extensive validation documentation support
  • Excellent for freezing applications
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 2 L
Capacity (English) 67.6 oz.
Color Clear
Diameter (English) Inner Neck 1.89 in.
Diameter (Metric) Inner Neck 48 mm
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
Material Polycarbonate
No. per Case 20/Cs.
No. per Pack 5/Pk.
Sterility Sterile
Temperature Range (Metric) -100°C to 100°C
Weight (English) 14.5 lb.
Closure Color White
Low Particulate <1/3 USP <788> (Ultralow particulate)
Certifications/Compliance USP Class VI, USP 661, FDA 21 CFR 177.1315
Closure Included Yes
Autoclavable Not Autoclavable
Mouth Narrow
Shape Square
Height (Metric) 265 mm
Product Type Media Bottle
Width (English) 19.75 in.
Closure Liner Silicone
Closure Material Polypropylene
Closure Size 38 mm
Diameter (Metric) 116 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.