Learn More
Thermo Scientific™ Cryoware Permanent Markers and Pens
GREENER_CHOICE
Catalog No. 1338252
Change view
Click to view available options
Color:
Assorted (Red, Blue, Green, Black)
Black
No. per Case:
10
24
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. per Case
13-382-52 Assorted (Red, Blue, Green, Black) 24
12-565-237 Black 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-382-52 Supplier Thermo Scientific™ Supplier No. 63130010
Only null left
Add to Cart
Add to Cart
Identify samples and storage boxes permanently with these cryoware permanent markers and pens.
Permanently notate samples and storage boxes with Thermo Scientific™ Cryoware Permanent Markers and Pens, which can easily withstand cold storage temperatures.
Features:
  • Nunc™ and Nalgene™ cryoware markers come in a variety of colors for permanent handwritten identification of samples
  • Use these markers by hand, or for higher throughput use them in the ID Scribe
TRUSTED_SUSTAINABILITY
Specifications
Product Type Permanent Marking Pen
Tip Style Extra Fine (Green is Fine)
Color Assorted (Red, Blue, Green, Black)
Description Cryoware Marker Set
No. per Case 24
No. per Pack 4
Quantity 4 Pk.