Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ LDPE, Wide-Mouth Carboy with Handle
GREENER_CHOICE

Catalog No. 0296160C
Click to view available options
Capacity (Metric):
10 L
15 L
20 L

For Use With: Store and transport solids, powders and liquids; Closure Material: Polypropylene; Color: Natural

Store and transport solids and powders with the Thermo Scientific™ Nalgene™ Wide-Mouth LDPE Carboys with Handles. Convenient large opening promotes ease of cleaning and dispensing and can accommodate overhead mixers while its wide-shoulder handles enable easy carrying and pouring, even with gloved hands. Carboy includes leakproof white polypropylene (PP) 100 mm screw closures.

  • Constructed of low density polyethylene
  • Graduated to contain in 1 gal. and 5 L increments for ease of use
  • Wide-mouth opening to accommodate overhead mixers and is convenient for dry materials such as solids and powders
  • Leakproof†
  • Not autoclavable
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Capacity (English) 4 gal.
Capacity (Metric) 15 L
Closure Material Polypropylene
Closure Size 100-415 mm
Color Natural
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Storage, Transportation ​
Material LDPE
No. per Case 6/Cs.
No. per Pack 1 Ea.
Product Type Carboy with Handle
Sterility Sterile
Volume (English) Brim 4.25 gal.
Volume (Metric) Brim 17 L
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.