Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Lined HDPE Serum Vial Closure for Continuous-Thread Vials: Nonsterile
GREENER_CHOICE

Catalog No. p-4526620
Click to view available options
Color:
Black
White

Eliminate the need for a rubber stopper while maintaining a leakproof seal with these lined, HDPE serum vial closures, for use with continuous thread PETG serum vials.

Eliminate the need for a rubber stopper, yet maintain a leakproof† seal with Thermo Scientific™ Nalgene™ Lined HDPE Serum Vial Closures for Continuous-Thread Vials. These nonsterile, high density polyethylene closures are for use with continuous-thread PETG serum vials.

Features:

  • Three-ply co-extruded liner (LDPE core between two facings of solid HDPE)
  • For use with nonsterile Nalgene PETG serum vials

Ordering Information:

Serum vial closures are bulk packed 2,000 per case.

TRUSTED_SUSTAINABILITY

Specifications

Closure Color White
Description Serum vial closures
Material (Liner) Three-ply co-extruded liner (LDPE core between two facing of solid HDPE)
Color White
Diameter (Metric) Outer 22.1 mm
Material HDPE
Sterility Non-sterile
No. per Case 2000
No. per Pack 2000
Product Line Nalgene
Closure Material HDPE
Closure Type Screw Cap
Height (Metric) 14.2 mm
Quantity 2000 Cs.
Certifications/Compliance ISO 13485:2003
Type Serum Vial Closure
Closure Size 20-415
For Use With (Equipment) Nonsterile Nalgene PETG Serum Vials
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.