Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Narrow-Mouth Opaque Amber HDPE Packaging Bottles without Closure: Bulk Pack
GREENER_CHOICE

Catalog No. p-4521833
Click to view available options
Capacity (Metric):
30 mL
125 mL
Closure Size:
20-415 mm
24-415 mm
This item is not returnable. View return policy

Catalog No. 03-313-86A

Supplier: Thermo Scientific™ 3620850001

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Protect light-sensitive reagents with these opaque, amber packaging bottles, which meet requirements of light-resistant containers per USP latest edition.

Protect valuable light-sensitive reagents with Thermo Scientific™ Nalgene™ Narrow-Mouth Opaque Amber HDPE Packaging Bottles. Packed without closures, you can match these bottles with various colored closures (Cat. No. 362150 series). These high-density, polyethylene bottles meet the requirements of light-resistant containers per USP latest edition.

  • Leakproof† with appropriate Nalgene closure

Ordering Information:

Bottles are bulk packed without closures. Order colored closures separately (Cat. No. 362150).

TRUSTED_SUSTAINABILITY

Specifications

Capacity 1 oz., 30 mL
Capacity (Metric) 30 mL
Capacity (English) 1 oz.
Color Opaque Amber
Description Narrow-Mouth Packaging Bottles
Diameter (Metric) Inner Neck 14 mm
Material HDPE
No. per Case 1000
No. per Pack 1000
Closure Color Opaque Amber
Neck Finish 20 mm
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Classs VI. Product meets the requirements for light-resistant containers per USP latest edition.
Closure Included Yes
Mouth Narrow
Shape Round
Height (Metric) 61 mm
Product Type Packer Bottle
Height (English) 2.4 in.
Closure Material Polypropylene
Closure Size 20-415 mm
Diameter (Metric) 34 mm
Closure Type Screw (Bulk Packed, Unassembled)
Through the Closure Dispensing No
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.