Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Polycarbonate Centrifuge Tubes w/Sealing Cap
GREENER_CHOICE

Catalog No. 055637A
Change view
Click to view available options
Capacity (Metric):
10 mL
16 mL
30 mL
50 mL
Length (Metric):
83.8 mm
102.6 mm
107.2 mm
114.8 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Length (Metric)
05-563-7A 10 mL 83.8 mm
05-563-7E 30 mL 102.6 mm
05-563-7G 50 mL 114.8 mm
05-563-7B 16 mL 107.2 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Autoclaving repeatedly will shorten the life of polycarbonate tubes. Tubes must be filled to at least 80% capacity for proper performance.
Catalog No. 05-563-7A Supplier Thermo Scientific™ Supplier No. 31380010
Add to Cart
Add to Cart
Autoclaving repeatedly will shorten the life of polycarbonate tubes. Tubes must be filled to at least 80% capacity for proper performance.

View contents easily using these strong, transparent polycarbonate centrifuge tubes, available with sealing caps for use up to 50,000 x g.

Easily view contents using strong, transparent Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Polycarbonate Centrifuge Tubes, equipped with sealing caps for leakproof† performance up to 50,000 x g.

Features:
  • Withstand temperatures from -135° to +135°C (-211° to +275°F)
  • Round bottom to fit high speed rotors
  • Can be used up to 50,000 x g in refrigerated or non-refrigerated centrifuges
  • Come with PP sealing caps with silicone gaskets for extra leak protection when spinning >10,000 x g or spinning hazardous materials
  • Autoclavable; transparent

Ordering Information:

Sealing caps also sold separately for 30 and 50mL sizes.

TRUSTED_SUSTAINABILITY

Specifications

Closure Material Polypropylene, red rubber
Capacity (Metric) 10 mL
Material Polycarbonate
No. per Case 50
No. per Pack 10
Sterility Non-sterile
Autoclavable Autoclavable
Shape Round
Length (Metric) 83.8 mm
Bottom Shape Round
Closure Type Screw Cap
Writing Area No
Diameter (Metric) 16.1 mm
Graduated No
Product Type Centrifuge Tube
Max. RCF 50,000 x g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.