Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Oak Ridge High-Speed PPCO Centrifuge Tubes with Sealing Cap
GSA_VAGREENER_CHOICE

Catalog No. 0556310G
Change view
Click to view available options
Capacity (Metric):
10 mL
16 mL
30 mL
50 mL
Length (Metric):
82.8 mm
102.1 mm
106.4 mm
113.5 mm
Diameter (Metric):
16.1 mm
18.1 mm
25.8 mm
28.8 mm
Max. RCF:
100,605 x g
50,000 x g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Length (Metric) Diameter (Metric) Max. RCF
05-563-10G 50 mL 113.5 mm 28.8 mm 100,605 x g
05-563-10A 10 mL 82.8 mm 16.1 mm 50,000 x g
05-563-10B 16 mL 106.4 mm 18.1 mm 50,000 x g
05-563-10E 30 mL 102.1 mm 25.8 mm 50,000 x g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 05-563-10G Supplier Thermo Scientific™ Supplier No. 31390050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Use these contact-clear high-speed PPCO centrifuge tubes with sealing caps to easily view the contents.

Spin with confidence with Thermo Scientific™ Nalgene™ Oak Ridge High-Speed PPCO Centrifuge Tubes with Sealing Caps. These tubes are chemically compatible with a wider variety of lab reagents than polycarbonate centrifuge tubes and are repeately autoclavable for extended performance.

Features

  • Greater chemical resistance than polycarbonate
  • Can be autoclaved repeatedly—last longer than polycarbonate tubes under conditions of repeated autoclaving
  • Can be used up to 50,000 x g in refrigerated or non-refrigerated centrifuges; 3139-0050 can be used up to 100,605 x g in the T29-8x50 rotor
  • Come with polypropylene sealing caps with silicone gaskets for extra leak protection when spinning >10,000 x g or spinning hazardous materials
  • Must be filled to 80% of total capacity when centrifuging
  • Autoclavable/leakproof†

Ordering Information:

Optional sealing-cap assemblies available for selected sizes. Use DS3131-0020 (for 30mL tube) and DS3131-0024 (for 50mL Tube). Actual brim capacity of 3139-0050 is 42mL.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Closure Material Polypropylene, red rubber
Capacity (Metric) 50 mL
Material Polypropylene Copolymer
No. per Case 50
No. per Pack 10
Sterility Non-sterile
Autoclavable Autoclavable
Shape Round
Length (Metric) 113.5 mm
Bottom Shape Round
Closure Type Screw Cap
Writing Area No
Diameter (Metric) 28.8 mm
Graduated No
Product Type Centrifuge Tube
Max. RCF 100,605 x g
Show More Show Less
Are there replacement O rings for Nalgene Oak Ridge High-Speed PPCO Centrifuge Tubes with Sealing Cap?

Replacement sealing-cap assemblies are available for selected sizes. We recommend Nalgene Centrifuge Tube Sealing Cap Assemblies (Cat. No. DS3131-0020) for 30 mL tubes and Nalgene Centrifuge Tube Sealing Cap Assemblies (Cat. No. DS3131-0024) for 50 mL tubes.
The actual brim capacity of the Nalgene Oak Ridge High-Speed PPCO Centrifuge Tubes with Sealing Cap is 42 mL, but it is designed to fit a standard rotor.

Are the Nalgene PPCO media bottles/tubes able to be irradiated?

PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.