Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles without Closure: Bulk Pack
GREENER_CHOICE

Catalog No. p-4521867
Click to view available options
Capacity (Metric):
4 mL
8 mL
15 mL
Neck Finish:
13-415 mm
20-415 mm

Safely store and ship light-sensitive, small-volume, diagnostic samples and valuable reagents with these opaque, amber HDPE bottles.

Safely store and ship light-sensitive, diagnostic samples and valuable reagents in small volumes with Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles, used in test kits and many small-volume applications. These opaque, amber, high-density polyethylene bottles are for use with light-sensitive reagents and are packed without closures so they can be matched with various colored closures (Cat. No. 362150 series).

  • Nonsterile
  • Durable, uniform walls resistant to splitting or puncturing
  • Leakproof† when assembled with closure (must be ordered separately)

Ordering Information:

Bottles are bulk packed without closures. Packed 2,000 per case. Order colored closures separately (Cat. No. 362150).

TRUSTED_SUSTAINABILITY

Specifications

Capacity 0.125 oz., 4 mL
Capacity (Metric) 4 mL
Capacity (English) 0.125 oz.
Color Opaque Amber
Description 3.4mL, 20-415 neck finish, bulk pack
Diameter (Metric) Inner Neck 8 mm
Material HDPE
No. per Case 2000
No. per Pack 2000
Sterility Non-sterile
Neck Finish 13-415 mm
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Classs VI. Product meets the requirements for light-resistant containers per USP latest edition.
Closure Included No
Graduated No
Mouth Narrow
Shape Round
Height (Metric) 41 mm
Product Type Sample Container
Width (English) 0.62 in.
Height (English) 1.61 in.
Closure Material Polypropylene
Closure Size 13-415 mm
Diameter (Metric) 16 mm
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.