Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ PETG Serum Vials with Continuous Thread: Nonsterile, Shrink-Wrapped Modules
GREENER_CHOICE

Catalog No. p-4526622
Click to view available options
Volume (Metric):
5 mL
10 mL
20 mL

Protect against leakage, breakage and contamination by using PETG nonsterile serum vials with continuous thread closures.

Protect against leakage, breakage, and contamination using Thermo Scientific™ Nalgene Nonsterile Serum Vials. Lighter than glass. Use with lined continuous thread closures (available separately).

Features:

  • PETG
  • Noncytotoxic
  • Nonpyrogenic
  • Packaged in trayless shrink-wrap modules
  • USP VI-compliant
  • Nonsterile
  • Leakproof† with Nalgene lined continuous-thread closures (available separately)

Ordering Information:

Serum vials are packed in trayless shrink-wrapped modules. Order closures separately (Cat. No. 312158).

TRUSTED_SUSTAINABILITY

Specifications

Description Serum Vials
Diameter (Metric) Inner 11 mm
Color Clear
Diameter (Metric) Outer 22 mm
Material PETG
Sterility Non-sterile
Volume (Metric) 5 mL
No. per Case 1656
No. per Pack 276
Volume (English) 1/6 oz.
Product Line Nalgene
Volume (Metric) Total 5 mL
Closure Type Screw Cap (Not Included)
Height (Metric) 44 mm
Quantity 276 Pk. , 1656 Cases
Certifications/Compliance ISO 13485:2003
Type Serum Vial
Closure Size 20-415
For Use With (Equipment) Lined Continuous Thread Closure
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.