Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Sealing Caps for Oak Ridge Centrifuge Tubes
GREENER_CHOICE

Catalog No. 055632D
Change view
Click to view available options
Closure Size:
20 mm
24 mm
58 mm
63 mm
For Use With (Equipment):
250 mL Tubes
28 and 30 mL Tubes
50 mL Tubes
500 mL and 1 L Tubes
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size For Use With (Equipment)
05-563-2D 58 mm 250 mL Tubes
05-563-2B 20 mm 28 and 30 mL Tubes
05-563-2C 24 mm 50 mL Tubes
05-563-2E 63 mm 500 mL and 1 L Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 05-563-2D Supplier Thermo Scientific™ Supplier No. DS31320058
Add to Cart
Add to Cart

Use these sealing caps to ensure leakproof service of centrifuge bottles at their recommended maximum rated speeds.

Ensure leakproof† service of Thermo Scientific™ Nalgene™ centrifuge bottles at their recommended maximum rated speeds by using Nalgene Sealing Caps for Oak Ridge Centrifuge Tubes. They are also recommended for use when spinning hazardous materials.

Compatibility:

  • DS3132-0058: for use with Nalgene Centrifuge Bottles 3140-0250 and 3140-0250
  • DS3132-0063: ror use with Nalgene Centrifuge Bottles 3140-0500, 3141-0500, 3120-1000, 3120-1010, 3122-1000 and 3122-1010
  • DS3132-0020: for use with Nalgene Oakridge tubes, 28ml and 30ml volumes
  • DS3132-0024: for use with Nalgene Oakridge tubes, 35ml and 50ml volumes

Note:

Before autoclaving, set closure on top of the container without engaging the threads.

TRUSTED_SUSTAINABILITY

Specifications

Closure Size 58 mm
Product Type Centrifuge Tube Cap
Autoclavable Autoclavable
Description Sealing Caps for Oak Ridge Centrifuge Tubes
Material Polypropylene, Rubber
No. per Pack 2
No. per Case 2
Product Line Nalgene
For Use With (Equipment) 250 mL Tubes

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.