Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Square Amber PETG Media Bottles with Closure: Nonsterile, Shrink-Wrapped Trays
GREENER_CHOICE

Catalog No. p-4521796
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
Closure Size:
20 mm
24 mm
38-430 mm

Safely store light-sensitive medias, buffers and sera in these square, amber PETG bottles.

Thermo Scientific™ Nalgene Square Amber PETG Media Bottles include HDPE closures. Translucent amber bottles meet USP light-resistant requirements and are ideal for medias, buffers and sera.

  • Heavy-walled, shatter resistant and durable
  • Space-saving square design
  • Molded in graduations
  • Nonsterile, non-pyrogenic, non-cytotoxic
  • Leakproof†, with drip-proof neck finish and molded-in neck ring
  • PETG provides excellent gas barrier, with transparency and reduced permeability to CO2 and O2
  • Amber bottle and closure provide protection for light sensitive reagents

Ordering Information:

Products are shrink-wrapped into trays, then packed in double PE lined cartons for cleanroom applications.

Recommended for:

For storage and shipping of light sensitive liquid media, buffers, and sera

Note:

† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 30 mL
Capacity (English) 1 oz.
Color Amber
Diameter (English) 1.5 in.
Diameter (English) Inner Neck 0.55 in.
Diameter (Metric) Inner Neck 14 mm
Material PETG
No. per Case 280
No. per Pack 40
Sterility Non-sterile
Certifications/Compliance PETG Media Bottles and HDPE closure materials meet current biological testing standards including USP Class VI.
Closure Included Yes
Graduated Yes
Mouth Wide
Shape Square
Height (Metric) 64 mm
Product Type Media Bottle
Height (English) 2.5 in.
Closure Material HDPE
Closure Size 20 mm
Diameter (Metric) 38 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.