Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Syringe Filters, Non-sterile
GREENER_CHOICE

Catalog No. 09740117
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
1.1 μm
Diameter (Metric):
4 mm
13 mm
25 mm
Material (Membrane):
Glass Fiber
Nylon
PTFE
SFCA
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Diameter (Metric) Material (Membrane)
09-740-117 0.2 μm 25 mm Nylon
09-740-101 0.2 μm 13 mm PTFE
09-740-102 0.45 μm 13 mm PTFE
09-740-103 1.1 μm 25 mm Glass Fiber
09-740-110 0.2 μm 25 mm SFCA
09-740-111 0.45 μm 25 mm SFCA
09-740-118 0.45 μm 25 mm Nylon
09-740-119 0.2 μm 25 mm PTFE
09-740-120 0.45 μm 25 mm PTFE
09-740-34C 0.2 μm 4 mm Nylon
09-740-34D 0.45 μm 4 mm Nylon
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 09-740-117 Supplier Thermo Scientific™ Supplier No. 7272020
Add to Cart
Add to Cart
This item is not returnable. View return policy

Filter sample volumes of 10 to 100mL with these nonsterile syringe filters featuring a variety of membranes and pore sizes for most laboratory applications.

Thermo Scientific™ Nalgene™ syringe filters are built to meet your lab’s clarification and sterilization needs. Our filters are available in a variety of sizes and membranes that can be easily used for both sterile and non-sterile laboratory applications. Whether your research necessitates prefiltration, sample preparation, sterile filtration, laboratory filtration, or gas venting, Nalgene syringe filters are lab essentials.

Key Features
  • 10 to 100mL sample size
  • Inlet: Female Luer-Lok™
  • Outlet: Male Luer slip
  • Polypropylene housing with ID color rings
  • Pore size and membrane type printed on unit

Surfactant-free Cellulose Acetate (SFCA)

  • Low protein binding, lower extractables than standard CA
  • Good for cell culture fluids and cellular solutions
  • Polypropylene housings with blue color rings

Cellulose Acetate (CA)

  • Some protein binding and extractables
  • For sterile filtration and clarification of: buffers, aqueous solutions and reagents
  • Polypropylene housings with red color rings

Nylon

  • Naturally hydrophilic, low extractables, some protein binding
  • Useful for alcohols, cryoprotectants such as DMSO and with aqueous/biological and weak solvents
  • Polypropylene housings with pink color rings
PTFE
  • Naturally hydrophobic
  • Excellent for HPLC/GC solvent filtration. Ideal for sterile venting of air or other gases during filtration
  • Polypropylene housings with white color rings

TRUSTED_SUSTAINABILITY

Specifications

Material (Membrane) Nylon
Pore Size 0.2 μm
Sterility Non-sterile
For Use With (Application) Filtration of Cryprotectants such as DMSO, aqueous biological solutions, alcoholic solutions and nonaqueous/HPLC/GC solvent applications
Green Features Fewer resources used and less waste
Housing Material Polypropylene (Housing)
Product Line Nalgene
Type Syringe Filter
Volume (Metric) 10 to 100 mL (Sample)
Diameter (Metric) 25 mm
Diameter (Metric) Membrane 25 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.