Learn More
Thermo Scientific™ Nalgene™ Syringe Filters, Non-sterile

Description
Thermo Scientific™ Nalgene™ syringe filters are built to meet your lab’s clarification and sterilization needs. Our filters are available in a variety of sizes and membranes that can be easily used for both sterile and non-sterile laboratory applications. Whether your research necessitates prefiltration, sample preparation, sterile filtration, laboratory filtration, or gas venting, Nalgene syringe filters are lab essentials.
- 10 to 100mL sample size
- Inlet: Female Luer-Lok™
- Outlet: Male Luer slip
- Polypropylene housing with ID color rings
- Pore size and membrane type printed on unit
Surfactant-free Cellulose Acetate (SFCA)
- Low protein binding, lower extractables than standard CA
- Good for cell culture fluids and cellular solutions
- Polypropylene housings with blue color rings
Cellulose Acetate (CA)
- Some protein binding and extractables
- For sterile filtration and clarification of: buffers, aqueous solutions and reagents
- Polypropylene housings with red color rings
Nylon
- Naturally hydrophilic, low extractables, some protein binding
- Useful for alcohols, cryoprotectants such as DMSO and with aqueous/biological and weak solvents
- Polypropylene housings with pink color rings
- Naturally hydrophobic
- Excellent for HPLC/GC solvent filtration. Ideal for sterile venting of air or other gases during filtration
- Polypropylene housings with white color rings
Specifications
Specifications
| Material (Membrane) | Nylon |
| Pore Size | 0.45 μm |
| Sterility | Non-sterile |
| For Use With (Application) | Filtration of Cryprotectants such as DMSO, aqueous biological solutions, alcoholic solutions and nonaqueous/HPLC/GC solvent applications |
| Green Features | Fewer resources used and less waste |
| Housing Material | Polypropylene (Housing) |
| Product Line | Nalgene |
| Type | Syringe Filter |
| Volume (Metric) | 10 to 100 mL (Sample) |
| Diameter (Metric) | 25 mm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.