Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer with Closure made with Tefzel™

Utilize these chemically inert, translucent and autoclavable funnels which feature a TFE stopcock assembly. Chemical-resistant, leakproof and requires no lubrication.

Supplier:  Thermo Scientific™ 43011000

 View more versions of this product

Catalog No. 10-437-25D

  • $1,390.65 / Case of 4

    Save 38%

    Reg : $2,240.00

    The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

Description

Description

Perform your liquid-liquid extractions with these chemically inert, translucent and autoclavable Thermo Scientific™ Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer and Closure made with Tefzel™ which feature a TFE stopcock assembly. TFE stopcock assembly has excellent chemical resistance, is leakproof† when used with provided Nalgene closure and does not require lubrication — can be chemically disinfected. More transparent than our polypropylene separatory funnels. Compatible with all common lab reagents and solvents. Won't chip, crack or shatter like glass separatory funnels.

Features:
  • Break-resistant and impervious to any chemical used in a separatory funnel
  • No phthalates to contaminate analyses
  • Useful in radiological and hazardous separations
  • Autoclavable/Leakproof
  • Stem length: 65mm

Includes:

Teflon™ FEP funnel, TFE stopcock and Tefzel™ ETFE screw closure

Ordering Information:

For replacement stopcock assembly for 125mL to 1L sizes, order Cat. No. 714301-1025. For replacement stopcock assembly for 2L size, order Cat. No. 714301-1050.

Note:

Funnel can be autoclaved; stopcock can be chemically sterilized. No stopcock lubrication needed.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1000 mL
371 mm
1/Pk.; 4/Cs.
Teflon FEP
Separatory Funnel FEP 1000mL; 53mm closure
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Teflon™ is a trademark of The Chemours Company FC, LLC used under license by Nalge Nunc International Corporation.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes.

Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.