Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalidixic acid, 99%

Catalog No. p-7027995
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 389-08-2 | C12H12N2O3 | 232.239 g/mol

Nalidixic acid is a synthetic 1,8-naphthyridine antimicrobial agent. It is also used as an inhibitor of bacterial DNA gyrase. It plays an important role in the treatment of urinary tract infections caused by susceptible gram-negative microorganisms, including the majority of E. Coli, Enterobacter species, Klebsiella species and Proteus species.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nalidixic acid is a synthetic 1,8-naphthyridine antimicrobial agent. It is also used as an inhibitor of bacterial DNA gyrase. It plays an important role in the treatment of urinary tract infections caused by susceptible gram-negative microorganisms, including the majority of E. Coli, Enterobacter species, Klebsiella species and Proteus species.

Solubility
Soluble in diethyl ether, acetone, methylene chloride, chloroform and methanol. Insoluble in water.

Notes
Light sensitive and air sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 389-08-2
Molecular Formula C12H12N2O3
Molecular Weight (g/mol) 232.239
MDL Number MFCD00006884
InChI Key MHWLWQUZZRMNGJ-UHFFFAOYSA-N
Synonym nalidixic acid, nalidixin, nevigramon, nalidixate, uronidix, innoxalon, nalidixan, nalitucsan, sicmylon, unaserus
PubChem CID 4421
ChEBI CHEBI:100147
IUPAC Name 1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid
SMILES CCN1C=C(C(=O)C2=C1N=C(C=C2)C)C(=O)O

Specifications

Melting Point 228°C to 230°C
Quantity 5 g
Beilstein 750515
Merck Index 14,6359
Solubility Information Soluble in diethyl ether,acetone,methylene chloride,chloroform and methanol. Insoluble in water.
Formula Weight 232.24
Percent Purity 99%
Chemical Name or Material Nalidixic acid
Hazard Category H317-H334-H336-H341
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
EINECSNumber 206-864-7
RTECSNumber QN2885000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.