Learn More
Invitrogen™ Nalidixic Acid Polyclonal Antibody

Description
Working Concentration: 1.25 μg/mL. Sensitivity: 10 ng/mL Nalidixic Acid produces 37% inhibition in a competitive ELISA employing Nalidixic Acid polyclonal antibody.
Specifications
Specifications
| Antigen | Nalidixic Acid |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.1 |
| Gene Alias | C12H12N2O3; Nalidixate; Nalidixin; Nevigramon; Unaserus |
| Host Species | Sheep |
| Immunogen | Nalidixic Acid-BTG. |
| Purification Method | Purified |
| Quantity | 1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.