Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NALP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100640
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100640 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100640 Supplier Invitrogen™ Supplier No. PA5100640
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NLRP1.

NALP1 is a member of the Ced-4 family of apoptosis proteins. Ced-family members contain a caspase recruitment domain (CARD) and are known to be key mediators of programmed cell death. The encoded protein contains a distinct N-terminal pyrin-like motif, which is possibly involved in protein-protein interactions. This protein interacts strongly with caspase 2 and weakly with caspase 9. Overexpression of this gene was demonstrated to induce apoptosis in cells. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene, but the biological validity of some variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NALP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NLRP1
Gene Accession No. Q9C000
Gene Alias CARD7; caspase recruitment domain protein; caspase recruitment domain protein 7; caspase recruitment domain-containing protein 7; CIDED; CLR17.1; death effector filament-forming Ced-4-like apoptosis protein; DEFCAP; DEFCAP-L/S; DKFZp586O1822; Gm14; Gm15; KIAA0926; NAC; nacht; NACHT, leucine rich repeat and PYD (pyrin domain) containing 1; NACHT, leucine rich repeat and PYD containing 1; NACHT, LRR and PYD containing protein 1; NACHT, LRR and PYD domains-containing protein 1; NACHT, LRR and PYD domains-containing protein 1a; NACHT/LRR/pyrin domain-containing protein 1; Nalp1; Nalp1a; NLR family protein 1; NLR family pyrin domain containing 1; NLR family, pyrin domain containing 1; NLR family, pyrin domain containing 1A; Nlrp1; Nlrp1a; Nucleotide-binding domain and caspase recruitment domain; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 1; PP1044; resistant anthrax lethal toxin; SLEV1; VAMAS1
Gene Symbols NLRP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human NLRP1(Accession Q9C000), corresponding to amino acid residues K229-T279.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22861
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.