Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NALP10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583488
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583488 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583488 Supplier Invitrogen™ Supplier No. PA583488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RARYFSSYFT DEKQADRAFD IVQKNDILYK ACQVPGICWV VCSWLQGQME RGKVVLETPR NSTDIFMAYV STFLPPDDDG GCSELSRHRV Highest antigen sequence indentity to the following orthologs: Rat - 67%, Mouse - 67%.

NALP proteins are cytoplasmic proteins that form a subfamily within the larger CATERPILLER family and are thought to play a crucial role in cell proliferation and reproduction. Like all other NALP family members, NALP10 has a C-terminal leucine-rich repeat (LRR) region, an N-terminal Pyrin domain (PYD) followed by a NACHT domain, and a NACHT-associated domain. It was initially identified as PYNOD, an Apaf-1-like protein that binds to ASC, caspase-1 and IL-1-b, inhibiting the autoprocessing of caspase-1, caspase-1-mediated IL-1-b processing, and ASC aggregation. NALP10 is thus a potent regulator of apoptosis and inflammation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NALP10
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NLRP10
Gene Accession No. Q86W26
Gene Alias 6430548I20Rik; CLR11.1; NACHT, leucine rich repeat and PYD containing 10; NACHT, LRR and PYD containing protein 10; NACHT, LRR and PYD domains-containing protein 10; NACHT/LRR/pyrin domain-containing protein 10; NALP10; Napl10; neuronal apoptosis inhibitor protein 10; NLR family pyrin domain containing 10; NLR family, pyrin domain containing 10; NLRP10; NOD8; Nucleotide-binding oligomerization domain protein 8; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 10; PAN5; Pynod
Gene Symbols NLRP10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NLRP10. Recombinant protein control fragment (Product #RP-97521).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338322
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.