Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ NALP12 Antibody Blocking Peptide

Click to view available options
Quantity:
0.1 mg
Specifications
Specifications
Gene ID (Entrez) | 91662 |
Species | Human |
Concentration | 1 mg/mL |
Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-56157 |
Gene Alias | FCAS2, Monarch-1, NACHT, leucine rich repeat and PYD containing 12, NACHT, LRR and PYD domains-containing protein 12, NALP12LRR and PYD containing protein 12, NLR family, pyrin domain containing 12, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 12, PYPAF7Monarch1, PYRIN-containing APAF1-like protein 7, Regulated by nitric oxide, RNO2 |
Gene Symbol | NLRP12 |
Quantity | 0.1 mg |
Research Category | Apoptosis |
Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction